Order Entry
Italy
ContactUsLinkComponent
Lacosamide Related Compounds, Reference Standard
Lacosamide Related Compounds, Reference Standard
Supplier:  USP
Certificates
Lacosamide Related Compounds, Reference Standard
Supplier:  USP

Specifications

  • CAS No.:
    1379592-19-4
  • Synonym(s):
    2-Amino-N-benzyl-3-methoxypropanamide oxalate
  • Description:
    Lacosamide  Related Compound D

Specifications

Frequently Bought Together