Order Entry
Italy
Orders LinkContactUsLinkComponent
Felodipine, Reference Standards

Felodipine, Reference Standards

Supplier:   USP
Felodipine, Reference Standards
Supplier:  USP
Certificates

2 Options Available Below

Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 1269389.
  • 1269390.
  • Description:
  • Felodipine
  • Felodipine Related Compound A
  • Synonym(s):
  • 3-Ethyl 5-methyl 4-(2,3-dichlorophenyl)-2,6-dimethylpyridine-3,5-dicarboxylate
  • CAS No.:
  • 72509-76-3
  • 96382-71-7
  • UN No.:
  • 3077

Specifications

Product Family Options

Product Information

  • Standards
    Pack type
    Pk
    Availability
    Price
  • 1269389.
    Felodipine
    Felodipine
    USP reference standard
    Vial
    300 mg
    Log in for availability
    Log in for availability
    1 * 300 mg
    1 * 300 mg - 354,00 €
    354,00 €
    1 * 300 mg
       354,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1269389.
    Description
    Felodipine
    Synonym(s)
    CAS No.
    72509-76-3
    UN No.
    3077
  • 1269390.
    Felodipine Related Compound A
    Felodipine Related Compound A
    USP reference standard
    Vial
    15 mg
    Log in for availability
    Log in for availability
    1 * 15 mg
    1 * 15 mg - 1 170,00 €
    1 170,00 €
    1 * 15 mg
       1 170,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1269390.
    Description
    Felodipine Related Compound A
    Synonym(s)
    3-Ethyl 5-methyl 4-(2,3-dichlorophenyl)-2,6-dimethylpyridine-3,5-dicarboxylate
    CAS No.
    96382-71-7
    UN No.
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.