Order Entry
Italy
ContactUsLinkComponent
Cevimeline Related Compounds, Reference Standard
Cevimeline Related Compounds, Reference Standard
Supplier:  USP
Certificates
Cevimeline Related Compounds, Reference Standard
Supplier:  USP

Specifications

  • CAS No.:
    1462862-74-3
  • Storage conditions:
    Refigerator
  • UN No.:
    2811
  • Synonym(s):
    (2RS,2'SR)-2'-Methyl-4-azaspiro{bicyclo[2.2.2]octane-2,5'-[1,3]oxathiolane} hydrochloride hemihydrate
  • Formula:
    C₁₀H₁₇NOS·HCl·₁/₂H₂O
  • Description:
    Cevimeline trans-Isomer
  • Melting point:
    201 - 203 °C
  • Physical form:
    Solid, crystaline powder

Specifications

Frequently Bought Together