Order Entry
Italy
Orders LinkContactUsLinkComponent
Allantoin, Reference Standard

Allantoin, Reference Standard

Allantoin, USP reference standard, 200 mg
Codice catalogo 1012939.
Nome del fornitore:   USP
Allantoin, Reference Standard
Codice catalogo 1012939.
Nome del fornitore:  USP
Numero CAS:  
Allantoin, Reference Standard

Allantoin, USP reference standard, 200 mg

Numero CAS:  
Price
369,00 €
Log in for availability
1 * 200 mg
Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Specifications

Specifications

  • Pk:
    200 mg
  • Standards:
    USP reference standard
  • Description:
    Allantoin
  • Cat. No.:
    1012939.
  • Formula:
    C₄H₆N₄O₃
  • Description:
    Allantoin
  • CAS No.:
    97-59-6

Specifications