Order Entry
Italy
ContactUsLinkComponent
2-Methyl-2-butanol ≥99%, GPR RECTAPUR®
 
 
2-Methyl-2-butanol ≥99%, GPR RECTAPUR®
  91242.320
 :  
undefined
 
 
2-Methyl-2-butanol ≥99%, GPR RECTAPUR®
  91242.320
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L
  • Seq:
    5096

 

 

  • Assay:
    Min. 99 %

 

 

 :
 :88.15 g/mol
 :13,07218
 :102 °C (1013 hPa)
 :–8.4 °C
 :0.806 g/cm³ (20 °C)
 :20 °C
 :Ambient
 :200-908-9
 :1105
 :3,II
 :GHS02
 :GHS07
 :75-85-4
 :C₅H₁₂O
 :MFCD00004478