Order Entry
Italy
ContactUsLinkComponent
Rotor package, Avanti JCF-Z
Rotor package, Avanti JCF-Z
Supplier:  Beckman Coulter
Certificates
undefined
Rotor package, Avanti JCF-Z
Supplier:  Beckman Coulter
Specifications
  • Catalog No:
  • 521-3596
  • 521-3912
  • Capacity:
  • Nominal capacity
    660 ml
  • Nominal capacity
    1250 ml
  • Max. RPM:
  • 20000
  • 20000