Order Entry
Italy
Orders LinkContactUsLinkComponent
Argatroban

Argatroban

Nome del fornitore:   Thermo Fisher Scientific
Argatroban
Nome del fornitore:  Thermo Fisher Scientific
Certificates

2 Options Available Below

Product Details & Documents
Synonyms:  MCI-9038, (2R,4R)-1-[(2S)-5-[(aminoiminomethyl)amino]-1-oxo-2-[[(1,2,3,4-tetrahydro-3-methyl-8-quinolinyl)sulphonyl]amino]pentyl]-4-methyl-2-piperidinecarboxylic acid, (2R,4R)-1-((2S)-5-Guanidino-2-(3-methyl-1,2,3,4-tetrahydroquinoline-8-sulphonamido)pentanoyl)-4-methylpiperidine-2-carboxylic acid
  • CAS Number:74863-84-6
  • Formula:C₂₃H₃₆N₆O₅S
  • MDL Number:MFCD23102419
  • Molecular Weight:508.64 g/mol
Specifications for Product Family

Specification Test Results

  • Water:
    =<4 %
  • Appearance (Form):
    Powder
  • HPLC:
    >=97.5 %
  • Appearance (Color):
    White to almost white
  • Infrared spectrum:
    Conforms
Product Family Options

Product Information

  • Pk
    Disponibilità
    Price
  • 250 mg
    Log in for availability
    Log in for availability
    1 * 250 mg
    1 * 250 mg - 347,00 €
    347,00 €
    1 * 250 mg
       347,00 €

  • 1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 949,00 €
    949,00 €
    1 * 1 g
       949,00 €