Order Entry
Italy
ContactUsLinkComponent
TCEP-HCl (Tris(2-carboxyethyl)phosphine hydrochloride), No-Weigh™, Pierce™
TCEP-HCl (Tris(2-carboxyethyl)phosphine hydrochloride), No-Weigh™, Pierce™
Catalog # PIERA35349
CAS Number:  
TCEP-HCl (Tris(2-carboxyethyl)phosphine hydrochloride), No-Weigh™, Pierce™
Catalog # PIERA35349
CAS Number:  

Specifications

  • Pk:
    10 mg
  • Seq:
    2275
  • Environmentally Preferable:
  • Pk Info:
    10 x 1 mg

Specifications

About this item

Synonyms: 3,3',3''-Phosphinetriyltripropanoic acid hydrochloride, TCEP, Tris(2-carboxyethyl)phosphine, hydrochloride
 :
 :286.65 g/mol
UN:3261
 :GHS05
Storage Temperature:Refrigerator
CAS Number:51805-45-9
Formula:C₉H₁₆ClO₆P
MDL Number:MFCD00145469
ADR:8 II