Order Entry
Italy
ContactUsLinkComponent
Glutaraldehyde 50% in water, GPR RECTAPUR®
Toxic
Corrosive
Health
Environment
Glutaraldehyde 50% in water, GPR RECTAPUR®
Catalog # 88452.230
Supplier:  VWR Chemicals
CAS Number:  
undefined
Toxic
Corrosive
Health
Environment
Glutaraldehyde 50% in water, GPR RECTAPUR®
Catalog # 88452.230
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pk:
    250 mL
  • Seq:
    2590

Specifications

Specification Test Results

  • Appearance:
    Clear colorless or yellow liquid
  • Assay (GC):
    47,5 - 52%
  • Color scale:
    max. 50 APHA
  • pH:
    3,2 - 4,2
  • Methanol:
    max. 0,3%

Specification Test Results

About this item

 :
 :100.12 g/mol
UN:2922
ADR:6.1(8),II
 :GHS06
 :GHS05
 :GHS08
 :GHS09
Storage Temperature:Ambient
CAS Number:111-30-8
Formula:C₅H₈O₂
MDL Number:MFCD00007025