Sulpho-LC-SPDP (Sulphosuccinimidyl 6-(3'-(2-pyridyldithio)propionamido)hexanoate) ≥90%, Pierce™
Catalog # PIER21650
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:50 mg
- Seq:2370
Specifications
About this item
:
:527.567 g/mol
Storage Temperature:Refrigerator
CAS Number:169751-10-4
Formula:C₁₈H₂₂N₃NaO₈S₃
MDL Number:MFCD00083162