Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Biocytin
Biocytin, EZ-Link™
Biocytin, EZ-Link™
Biocytin, EZ-Link™
Catalog # PIER28022
Supplier:
Thermo Fisher Scientific
CAS Number:
Expand image
Biocytin, EZ-Link™
Catalog # PIER28022
Supplier:
Thermo Fisher Scientific
CAS Number:
Specifications
Pk:
100 mg
Seq:
2440
Specifications
About this item
Product Details
:
:
372.484 g/mol
CAS Number:
576-19-2
Formula:
C₁₆H₂₈N₄O₄S
MDL Number:
MFCD00077319
Product Details