Order Entry
Italy
ContactUsLinkComponent
5,5'-Diallyl-2,2'-dihydroxybiphenyl (Magnolol)
Corrosive
Irritant
5,5'-Diallyl-2,2'-dihydroxybiphenyl (Magnolol)
Catalog # ACRO465512500
CAS Number:  
Corrosive
Irritant
5,5'-Diallyl-2,2'-dihydroxybiphenyl (Magnolol)
Catalog # ACRO465512500
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

Specification Test Results

  • Appearance:
    White to yellow to beige powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    99°C to 103°C
  • Water:
    ≤ 1%

Specification Test Results

About this item

 :
Melting Pt:101°C…102°C
 :266.34 g/mol
 :GHS05
 :GHS07
Storage Temperature:Refrigerator
CAS Number:528-43-8
Formula:C₁₈H₁₈O₂
MDL Number:MFCD00016658