Order Entry
Italy
Orders LinkContactUsLinkComponent
MPC series plastic couplings
MPC series plastic couplings
Nome del fornitore:   CPC COLDER
MPC series plastic couplings
Nome del fornitore:  CPC COLDER
Certificates
MPC series plastic couplings
14 Options Available Below
Product Details & Documents
Couplings allow rapid disconnect and reconnect with one hand and without the need for tools.
Couplings are available in polysulphone (amber tint) or polycarbonate (purple tint) construction. Coupling inserts include seals constructed of clear silicone. All products meet USP Class VI criteria.
They feature an ergonomic thumb latch and an audible click when connected, assuring the user of a secure connection. Couplings can be rotated when connected, reducing the inconvenience of tube kinks. These items are sterilisable and reusable, yet economical enough to allow disposability.
Certifications: All products meet USP Class VI criteria and feature BSE-/TSE-free materials (ADCF).
Specifications for Product Family
Specifications
  • Catalog No:
  • 212-0801
  • 212-0805
  • 212-0804
  • 212-0803
  • 212-0802
  • 212-0806
  • 212-0808
  • 212-0807
  • 212-0812
  • 212-0811
  • 212-0810
  • 212-0809
  • 212-0814
  • 212-0813
  • Max. operating pressure:
  • Vacuum...4,2 bar
  • Vacuum...4,2 bar
  • Vacuum...4,2 bar
  • Vacuum...4,2 bar
  • Vacuum...4,2 bar
  • Vacuum...4,2 bar
  • Vacuum...4,2 bar
  • Vacuum...4,2 bar
  • Vacuum...4,2 bar
  • Vacuum...4,2 bar
  • Vacuum...4,2 bar
  • Vacuum...4,2 bar
  • Vacuum...4,2 bar
  • Vacuum...4,2 bar
  • Operating temperature:
  • Polycarbonate –40...+121 °C, polysulphone –40...+148,9 °C
  • Polycarbonate –40...+121 °C, polysulphone –40...+148,9 °C
  • Polycarbonate –40...+121 °C, polysulphone –40...+148,9 °C
  • Polycarbonate –40...+121 °C, polysulphone –40...+148,9 °C
  • Polycarbonate –40...+121 °C, polysulphone –40...+148,9 °C
  • Polycarbonate –40...+121 °C, polysulphone –40...+148,9 °C
  • Polycarbonate –40...+121 °C, polysulphone –40...+148,9 °C
  • Polycarbonate –40...+121 °C, polysulphone –40...+148,9 °C
  • Polycarbonate –40...+121 °C, polysulphone –40...+148,9 °C
  • Polycarbonate –40...+121 °C, polysulphone –40...+148,9 °C
  • Polycarbonate –40...+121 °C, polysulphone –40...+148,9 °C
  • Polycarbonate –40...+121 °C, polysulphone –40...+148,9 °C
  • Polycarbonate –40...+121 °C, polysulphone –40...+148,9 °C
  • Polycarbonate –40...+121 °C, polysulphone –40...+148,9 °C
Specifications
Product Family Options
Coupling bodies
Product Information
  • Description
    Tubing int.Ø
    Disponibilità
    Price
  • Polycarbonate in-line hose barb, non-valved
    6,4 mm
    Log in for availability
    Log in for availability
    1 * 250 pezzi
    1 * 250 pezzi - 569,00 €
    569,00 €
    1 * 250 pezzi
       569,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    212-0801
    Max. operating pressure
    Vacuum...4,2 bar
    Operating temperature
    Polycarbonate –40...+121 °C, polysulphone –40...+148,9 °C
  • Polycarbonate in-line hose barb, non-valved
    9,5 mm
    Log in for availability
    Log in for availability
    1 * 25 pezzi
    1 * 25 pezzi - 78,70 €
    78,70 €
    1 * 25 pezzi
       78,70 €
    Specifications:
    More
    Specifications
    Cat. No.
    212-0805
    Max. operating pressure
    Vacuum...4,2 bar
    Operating temperature
    Polycarbonate –40...+121 °C, polysulphone –40...+148,9 °C
  • Polycarbonate in-line hose barb, non-valved
    9,5 mm
    Log in for availability
    Log in for availability
    1 * 250 pezzi
    1 * 250 pezzi - 569,00 €
    569,00 €
    1 * 250 pezzi
       569,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    212-0804
    Max. operating pressure
    Vacuum...4,2 bar
    Operating temperature
    Polycarbonate –40...+121 °C, polysulphone –40...+148,9 °C
  • Polysulphone in-line hose barb, non-valved
    6,4 mm
    Log in for availability
    Log in for availability
    1 * 25 pezzi
    1 * 25 pezzi - 131,00 €
    131,00 €
    1 * 25 pezzi
       131,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    212-0803
    Max. operating pressure
    Vacuum...4,2 bar
    Operating temperature
    Polycarbonate –40...+121 °C, polysulphone –40...+148,9 °C
  • Polysulphone in-line hose barb, non-valved
    6,4 mm
    Log in for availability
    Log in for availability
    1 * 250 pezzi
    1 * 250 pezzi - 946,00 €
    946,00 €
    1 * 250 pezzi
       946,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    212-0802
    Max. operating pressure
    Vacuum...4,2 bar
    Operating temperature
    Polycarbonate –40...+121 °C, polysulphone –40...+148,9 °C
  • Polysulphone in-line hose barb, non-valved
    9,5 mm
    Log in for availability
    Log in for availability
    1 * 250 pezzi
    1 * 250 pezzi - 946,00 €
    946,00 €
    1 * 250 pezzi
       946,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    212-0806
    Max. operating pressure
    Vacuum...4,2 bar
    Operating temperature
    Polycarbonate –40...+121 °C, polysulphone –40...+148,9 °C
Coupling inserts
Product Information
  • Description
    Tubing int.Ø
    Disponibilità
    Price
  • Polycarbonate, silicone seal, non-valved
    6,4 mm
    Log in for availability
    Log in for availability
    1 * 25 pezzi
    1 * 25 pezzi - 78,70 €
    78,70 €
    1 * 25 pezzi
       78,70 €
    Specifications:
    More
    Specifications
    Cat. No.
    212-0808
    Max. operating pressure
    Vacuum...4,2 bar
    Operating temperature
    Polycarbonate –40...+121 °C, polysulphone –40...+148,9 °C
  • Polycarbonate, silicone seal, non-valved
    6,4 mm
    Log in for availability
    Log in for availability
    1 * 250 pezzi
    1 * 250 pezzi - 569,00 €
    569,00 €
    1 * 250 pezzi
       569,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    212-0807
    Max. operating pressure
    Vacuum...4,2 bar
    Operating temperature
    Polycarbonate –40...+121 °C, polysulphone –40...+148,9 °C
  • Polycarbonate, silicone seal, non-valved
    9,5 mm
    Log in for availability
    Log in for availability
    1 * 25 pezzi
    1 * 25 pezzi - 78,70 €
    78,70 €
    1 * 25 pezzi
       78,70 €
    Specifications:
    More
    Specifications
    Cat. No.
    212-0812
    Max. operating pressure
    Vacuum...4,2 bar
    Operating temperature
    Polycarbonate –40...+121 °C, polysulphone –40...+148,9 °C
  • Polycarbonate, silicone seal, non-valved
    9,5 mm
    Log in for availability
    Log in for availability
    1 * 250 pezzi
    1 * 250 pezzi - 569,00 €
    569,00 €
    1 * 250 pezzi
       569,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    212-0811
    Max. operating pressure
    Vacuum...4,2 bar
    Operating temperature
    Polycarbonate –40...+121 °C, polysulphone –40...+148,9 °C
  • Polysulphone, silicone seal, non-valved
    6,4 mm
    Log in for availability
    Log in for availability
    1 * 25 pezzi
    1 * 25 pezzi - 128,00 €
    128,00 €
    1 * 25 pezzi
       128,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    212-0810
    Max. operating pressure
    Vacuum...4,2 bar
    Operating temperature
    Polycarbonate –40...+121 °C, polysulphone –40...+148,9 °C
  • Polysulphone, silicone seal, non-valved
    6,4 mm
    Log in for availability
    Log in for availability
    1 * 250 pezzi
    1 * 250 pezzi - 924,00 €
    924,00 €
    1 * 250 pezzi
       924,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    212-0809
    Max. operating pressure
    Vacuum...4,2 bar
    Operating temperature
    Polycarbonate –40...+121 °C, polysulphone –40...+148,9 °C
  • Polysulphone, silicone seal, non-valved
    9,5 mm
    Log in for availability
    Log in for availability
    1 * 25 pezzi
    1 * 25 pezzi - 128,00 €
    128,00 €
    1 * 25 pezzi
       128,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    212-0814
    Max. operating pressure
    Vacuum...4,2 bar
    Operating temperature
    Polycarbonate –40...+121 °C, polysulphone –40...+148,9 °C
  • Polysulphone, silicone seal, non-valved
    9,5 mm
    Log in for availability
    Log in for availability
    1 * 250 pezzi
    1 * 250 pezzi - 924,00 €
    924,00 €
    1 * 250 pezzi
       924,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    212-0813
    Max. operating pressure
    Vacuum...4,2 bar
    Operating temperature
    Polycarbonate –40...+121 °C, polysulphone –40...+148,9 °C