Anti-LCMT1 Mouse Monoclonal Antibody [clone: 2C9]
Catalog # NOVUNBP2-00923
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Leucine carboxyl methyltransferase 1
- Antigen symbol:LCMT1
- Clonality:Monoclonal
- Clone:2C9
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Primates,Human,Rat,Mouse
- Western blot:Yes
- Gene ID:51451
- Storage buffer:PBS (pH 7.3), 1.0% BSA and 50% Glycerol
- Storage temperature:Store at 4 C short term. Aliquot and store at -20 C long term. Avoid freeze-thaw cycles.
- Immunogen:Full length human recombinant protein of human LCMT1(NP_057393) produced in HEK293T cell.
- Purification:Immunogen affinity purified
- Size:100 μl
- Pk:0,1 mL
Specifications
About this item
Mouse Monoclonal LCMT1 Antibody (2C9). Tested Applications: Western Blot, Immunohistochemistry. Tested Reactivity: Human, Mouse, Rat, Primate.
Type: Primary
Antigen: LCMT1
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human