Order Entry
Italy
ContactUsLinkComponent
Anti-CD59 mouse monoclonal antibody [clone: MEM-43]
Anti-CD59 mouse monoclonal antibody [clone: MEM-43]
Catalog # NOVUNB500-330
Supplier:  Novus Biologicals
Anti-CD59 mouse monoclonal antibody [clone: MEM-43]
Catalog # NOVUNB500-330
Supplier:  Novus Biologicals

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    CD59
  • Clonality:
    Monoclonal
  • Clone:
    MEM-43
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG2a
  • Reactivity:
    Human
  • Neutralisation:
    Yes
  • Gene ID:
    966
  • Antigen synonyms:
    CD59 antigen|FLJ38134|membrane attack complex (MAC) inhibition factor|MIRL|Ly-6-like protein|MAC-inhibitory protein|HRF-20|MEM43|1F5|MACIF|MIN3|protectin|EJ16|EL32|MIC11MSK21|MGC2354|HRF20|p18-20|surface anitgen recognized by monoclonal 16.3A5|Membrane inhibitor of reactive lysis|EL32 and G344)|MIN1|lymphocytic antigen CD59/MEM43|CD59 molecule|16.3A5|MEM43 antigen|20 kDa homologous restriction factor|MIN2|complement regulatory protein|CD59 glycoprotein|1F5 antigen|CD59 antigen p18-20 (antigen identified by monoclonal antibodies 16.3A5|human leukocyte antigen MIC11|Membrane attack complex inhibition factor|MAC-IP|T cell-activating protein|G344|EJ30|FLJ92039
  • Storage buffer:
    PBS (pH 7,4)
  • Storage temperature:
    Store at 4 °C. Do not freeze.
  • Concentration:
    1 mg/ml
  • Immunogen:
    Thymocytes and T lymphocytes (NM_000611.5).
  • Purification:
    Protein A purified
  • Pk:
    0,1 mg

Specifications

About this item

The CD59 Antibody (MEM-43) from Novus Biologicals is a mouse monoclonal antibody to CD59. This antibody reacts with human. The CD59 Antibody (MEM-43) has been validated for the following applications: Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Blocking / Neutralising, CyTOF-ready.

Type: Primary
Antigen: CD59
Clonality: Monoclonal
Clone: MEM-43
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human