Anti-CUL-3 Rabbit polyclonal antibody
NOVUNB100-58788
- Antibody type:Primary
- Antigen name:Cullin 3
- Antigen symbol:CUL-3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Gene ID:8452
- Antigen synonyms:CUL-3|KIAA0617|cullin-3|cullin 3
- Storage buffer:TBS and 0.1% BSA
- Storage temperature:Store at 4 °C. Do not freeze.
- Concentration:0,2 mg/ml
- Immunogen:The immunogen recognized by this antibody maps to a region between residue 650 and 700 of human cullin 3 using the numbering given in entry NP_003581.1 (GeneID 8452).
- Purification:Immunogen affinity purified
- Pk:0,1 mL
The Cullin 3 Antibody from Novus Biologicals is a rabbit polyclonal antibody to Cullin 3. This antibody reacts with human, mouse. The Cullin 3 Antibody has been validated for the following applications: Western Blot, Immunoprecipitation, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: CUL-3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse