Order Entry
Italy
ContactUsLinkComponent
Diethanolamine 98.5-101.0%, GPR RECTAPUR®
Corrosive
Health
Irritant
Diethanolamine 98.5-101.0%, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates
undefined
Corrosive
Health
Irritant
Diethanolamine 98.5-101.0%, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay (calculated on anhydrous) :
    98.5 - 101.0 %
  • Identification (IR) Passes test:
    Passes test
  • Melting point (lower value):
    ≥ 25 °C
  • Melting point (upper value) :
    ≤ 29 °C
  • Water :
    ≤ 0.15 %

Specification Test Results

Related Information

Frequently Bought Together