GF Signet® Manual Diaphragm Valves, PVC
Optimized valve body provides double the CV values.
- Two-color positition indicator on handwheel
- Elimination of bolts removes corrosive weak points
- Mount vertically or horizontally
- PVC body and EPDM diaphragm
These GF Signet® manual diaphragm valves are constructed with a PVC body and EPDM diaphragm. Valves offer a maximum temperature of 150 °F (65 °C) and feature true union socket connections.
- Environmentally Preferable:
- Catalog No:
- MFLX07390-50
- MFLX07390-54
- MFLX07390-56
- Max. temperature:
- 65,5 °C (150 °F)
- 65,5 °C (150 °F)
- 65,5 °C (150 °F)
- Wetted materials:
- PVC, EPDM
- PVC, EPDM
- PVC, EPDM
- Working pressure:
- 150 psi
- 150 psi
- 150 psi
Frequently Bought Together