Enterobacter cloacae WDCM 00083 Vitroids™, Certified Reference Material, Suitable for Microbiology, Supelco®
Catalog # VT000834-10EA
Supplier: Merck
CAS Number:
Specifications
- Pk:10
- Description:Enterobacter cloacae WDCM 00083 Vitroids™, 130 - 300 CFU mean value range
- Environmentally Preferable:
- M-Clarity quality level:MQ300
- Storage temperature:–20 °C
- Product type:Hydrosoluble discs
Specifications
About this item
Derived from CECT® strains.
Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025.