Order Entry
Italy
Orders LinkContactUsLinkComponent
Tags
Tags
Supplier:   GA
Tags
Supplier:  GA
Certificates
Tags
5 Options Available Below
Product Details & Documents
These tags are used for identification of metal racks and other containers for long-term storage in liquid nitrogen and ultra-low temperature freezers.
  • Suitable temperature ranges from –196 to +80 °C
Labels can be printed with all major brands of thermal transfer barcode printers (SATO, Zebra, Intermec, Printronix, Datamax-O'Neil, CAB, and more) or inscribed with permanent markers such as our cryo-markers.
Colours available:
Yellow.
Assorted: Pink, Blue, Yellow.
Assorted 2: Purple, Green, Orange.
Assorted 3: Purple, Green, Orange, Pink, Blue, Yellow.
Delivery information: Includes 12 plastic Cryo-Cards™ with connection hooks and thermal transfer cryo labels.
Product Family Options
Product Information
  • Material
    Colour
    Dimensions
    Packed
    Availability
    Price
  • GAINCID-02WH
    Cryogenic tags
    Cryogenic tags
    Plastic
    White
    54,1×30 mm
    12 tags/pack
    Log in for availability
    Log in for availability
    1 * 12 pezzi
    1 * 12 pezzi - 28,30 €
    28,30 €
    1 * 12 pezzi
       28,30 €
  • GAINCIDB-02YE
    Cryogenic tags
    Cryogenic tags
    Plastic
    Yellow
    54,1×30 mm
    12 tags/pack
    Log in for availability
    Log in for availability
    1 * 12 pezzi
    1 * 12 pezzi - 28,30 €
    28,30 €
    1 * 12 pezzi
       28,30 €
    Specifications:
    New Product
    New Product
  • GAINCIDB-02A
    Cryogenic tags; Assorted: Pink, Blue, Yellow
    Cryogenic tags; Assorted: Pink, Blue, Yellow
    Plastic
    Assorted
    54,1×30 mm
    12 tags/pack
    Log in for availability
    Log in for availability
    1 * 12 pezzi
    1 * 12 pezzi - 28,30 €
    28,30 €
    1 * 12 pezzi
       28,30 €
    Specifications:
    New Product
    New Product
  • GAINCIDB-02A2
    Cryogenic tags; Assorted 2: Purple, Green, Orange
    Cryogenic tags; Assorted 2: Purple, Green, Orange
    Plastic
    Assorted
    54,1×30 mm
    12 tags/pack
    Log in for availability
    Log in for availability
    1 * 12 pezzi
    1 * 12 pezzi - 28,30 €
    28,30 €
    1 * 12 pezzi
       28,30 €
    Specifications:
    New Product
    New Product
  • GAINCIDB-02A3
    Cryogenic tags; Assorted 3: Purple, Green, Orange, Pink, Blue, Yellow
    Cryogenic tags; Assorted 3: Purple, Green, Orange, Pink, Blue, Yellow
    Plastic
    Assorted
    54,1×30 mm
    12 tags/pack
    Log in for availability
    Log in for availability
    1 * 12 pezzi
    1 * 12 pezzi - 28,30 €
    28,30 €
    1 * 12 pezzi
       28,30 €
    Specifications:
    New Product
    New Product