Order Entry
Italy
Orders LinkContactUsLinkComponent
 

PlusOne™ Repel-Silane ES, colourless liquid

500 mL
Catalog # 17-1332-01
Flammable
Health
Irritant
Supplier:   Cytiva
PlusOne™ Repel-Silane ES, colourless liquid
Catalog # 17-1332-01
Supplier:  Cytiva
CAS Number:  
PlusOne™ Repel-Silane ES, colourless liquid
PlusOne™ Repel-Silane ES, colourless liquid

500 mL

CAS Number:  
Price
371,00 €
Log in for availability
1 * 500 mL
Product Details & Documents

Repel-Silane ES is a 2% solution of dimethyldichlorosilane dissolved in octamethylcyclooctasilane.

  • Used in IEF, PAGE and Sequencing
  • Soluble in cold water and hot water

It is used to inhibit sticking of polyacrylamide gels, agarose gels, and nucleic acids to glass surfaces.

  • Flash Pt:56 °C (132.8 °F) (closed cup)
  • Storage Temperature:Ambient
  • UN:1993
  • ADR:3,III
Specifications

Specifications

  • Pk:
    500 mL

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.