Order Entry
Italy
Orders LinkContactUsLinkComponent
Corning® Trypsin

Corning® Trypsin

100 ml, 6
Catalog # MDTC25-054-CI
Supplier:   Corning
Corning® Trypsin
Catalog # MDTC25-054-CI
Supplier:  Corning
CAS Number:  
Corning® Trypsin
Corning® Trypsin

100 ml, 6

CAS Number:  
Price
211,00 €
Log in for availability
1 * 6 pezzi
Product Details & Documents

Trypsin is used to enzymatically release adherent cells from tissue culture plates for passaging

  • Trypsin 10X 2,5% Trypsin in HBSS without calcium, magnesium, and phenol red Porcine Parvovirus Tested

Divalent cations such as calcium and magnesium, which are often present in the cell culture environment, inhibit this action. EDTA sequesters these ions and thus enhances the efficacy of trypsin.

Documents

Specifications

Specifications

  • Pk:
    6
  • Pk Info:
    100 ml
  • Cat. No.:
    MDTC25-054-CI
  • Storage conditions:
    Frozen
  • Formulation:
    Trypsin 10X, 2,5% Trypsin in HBSS without Calcium, Magnesium, and Phenol Red
  • Enzyme name:
    Trypsin
  • Shipping conditions:
    Frozen

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.