Order Entry
Italy
Orders LinkContactUsLinkComponent
Amphotericin B 250 µg/ml

Amphotericin B 250 µg/ml

6×50 ml, 6
Catalog # MDTC30-003-CF
Supplier:   Corning
Amphotericin B 250 µg/ml
Catalog # MDTC30-003-CF
Supplier:  Corning
CAS Number:  1397-89-3
Amphotericin B 250 µg/ml
Amphotericin B 250 µg/ml

6×50 ml, 6

CAS Number:  1397-89-3
Price
161,00 €
Log in for availability
1 * 6 pezzi
Product Details & Documents

This antimycotic interferes with fungal membrane permeability by forming channels in cell membranes, causing small molecules to leak from the cell. It is recommended for use in cell culture applications at 10 ml/L and is stable at 37 ºC for 3 days.

Packaging: Shipped frozen.

Synonyms:  (1R,3S,5R,6R,9R,11R,15S,16R,17R,18S,19E,21E,23E,25E,27E,29E,31E,33R,35S,36R,37S)-33-(((2R,3S,4S,5S,6R)-4-Amino-3,5-dihydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)-1,3,5,6,9,11,17,37-octahydroxy-15,16,18-trimethyl-13-oxo-14,39-dioxabicyclo[33.3.1]nonatriaconta-19,21,23,25,27,29,31-heptaene-36-carboxylic acid
  • CAS Number:1397-89-3
  • Formula:C₄₇H₇₃NO₁₇
  • MDL Number:MFCD00877763
  • Molecular Weight:924.08 g/mol
  • Storage Temperature:Freezer

Documents

Specifications

Specifications

  • Pk:
    6
  • Pk Info:
    6×50 ml

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.