Micro-dispensers, Piccolo 1 and 2
: VITLAB
For dispensing tiny quantities of liquids in all areas of biochemical and medical research.
- Reset mechanism refills the volume automatically
- Disposable tips are unnecessary
- Can be operated with only one hand
- Use the thumb to depress the volume dispensing button, just as with a pipette
- Discharge tube can be rotated over 360° so that it is always optimally situated with respect to the bottle label
The Piccolo are designed for serial dispensing operations. In order to guarantee high resistance to chemicals and a broad range of applications, only high-quality materials, such as PTFE, PFA, ETFE, FEP, borosilicate glass, and platinum-iridium come in contact with the media.
: Supplied with mounting tools and an instruction manual.
Frequently Bought Together