Order Entry
Italy
ContactUsLinkComponent
Dexamethasone 100 mM
Dexamethasone 100 mM
Catalog # API-04
Supplier:  G-Biosciences
CAS Number:  
undefined
Dexamethasone 100 mM
Catalog # API-04
Supplier:  G-Biosciences
CAS Number:  

Specifications

  • Pk:
    1 mL
  • Seq:
    2070

Specifications

About this item

 :
 :392.47 g/mol
CAS Number:50-02-2
Formula:C₂₂H₂₉FO₅
MDL Number:MFCD00064136