Order Entry
Italy
Orders LinkContactUsLinkComponent
BM-2 orchid medium
BM-2 orchid medium
Supplier:   Apollo Scientific
BM-2 orchid medium
Supplier:  Apollo Scientific
Certificates
BM-2 orchid medium
2 Options Available Below
Product Details & Documents
Contains macro/micronutrients, vitamins and plantgrowth regulators required for orchid culturePlant tissue culture testedFull specification given in appendix.
  • MFCD07370631
Product Family Options
Product Information
  • Pk
    Availability
    Price
  • APOSPMB142A-10X1L
    BM-2 orchid medium 10×1 L
    BM-2 orchid medium 10×1 L
    10
    Log in for availability
    Log in for availability
    1 * 10 pezzi
    1 * 10 pezzi
  • APOSPMB142N-10L
    BM-2 orchid medium 10 L
    BM-2 orchid medium 10 L
    10 L
    Log in for availability
    Log in for availability
    1 * 10 L
    1 * 10 L - 21,80 €
    21,80 €
    1 * 10 L
       21,80 €