Vacuum degassers
HEWL5042-8922
: Agilent
- Description:Micro vacuum degasser tubing kit
- Seq:01
- Pk:1 KIT
A vacuum degasser is recommended for maximum sensitivity in the low UV wavelength range; high injection precision; high retention time reproducibility; and flow rates below 0,5 ml/min.
- To generally clean the vacuum degasser tubing, flush the system with isopropanol
- Flush the degasser with water after using buffers
Speed solvent changes by drawing solvent through the degasser and tubing with syringe adapter kit.