Order Entry
Italy
Orders LinkContactUsLinkComponent
VWR®, Cable Ties, Polyamide 6.6

VWR®, Cable Ties, Polyamide 6.6

VWR®, Cable Ties, Polyamide 6.6
Supplier:  VWR Collection
Certificates
VWR®, Cable Ties, Polyamide 6.6
VWR®, Cable Ties, Polyamide 6.6

16 Options Available Below

Product Details & Documents

Pull-tight cable ties are available in different lengths ranging from 100 to 370 mm. All cable ties are made from polyamide 6.6 material.

  • High resistance to bases, oils, greases, and oil derivatives
  • Limited resistance to acids
  • Not resistant to phenols or chloride solvents
  • Operating temperature: −40 up to 85 °C
  • Installation temperature: −10 up to 60 °C
  • Melting point: 260 °C
  • Flammability class V2 acc- a UL 94

Easy to use in a wide set of applications, cable ties provide secure fastening solutions. A variety of colours allows users to identify the right cable tie for the most appropriate use. Effective and long lasting closure system is activated by simply inserting the tail into the tie head and pulling to tighten around the target object.

Product Family Options

Product Information

  • Colour
    Dimensions
    Availability
    Price
  • 129-1607
    Cable ties, polyamide 6.6, resistance: 8
    Cable ties, polyamide 6.6, resistance: 8
    Black
    100×2,5 mm
    Log in for availability
    Log in for availability
    1 * 100 pezzi
    1 * 100 pezzi - 14,70 €
    14,70 €
    1 * 100 pezzi
       14,70 €

  • 129-1608
    Cable ties, polyamide 6.6, resistance: 18
    Cable ties, polyamide 6.6, resistance: 18
    Black
    150×3,6 mm
    Log in for availability
    Log in for availability
    1 * 100 pezzi
    1 * 100 pezzi - 18,20 €
    18,20 €
    1 * 100 pezzi
       18,20 €

  • 129-1609
    Cable ties, polyamide 6.6, resistance: 22
    Cable ties, polyamide 6.6, resistance: 22
    Black
    200×4,8 mm
    Log in for availability
    Log in for availability
    1 * 100 pezzi
    1 * 100 pezzi - 22,40 €
    22,40 €
    1 * 100 pezzi
       22,40 €

  • 129-1610
    Cable ties, polyamide 6.6, resistance: 22
    Cable ties, polyamide 6.6, resistance: 22
    Black
    300×4,8 mm
    Log in for availability
    Log in for availability
    1 * 100 pezzi
    1 * 100 pezzi - 37,70 €
    37,70 €
    1 * 100 pezzi
       37,70 €

  • 129-1611
    Cable ties, polyamide 6.6, resistance: 22
    Cable ties, polyamide 6.6, resistance: 22
    Black
    370×4,8 mm
    Log in for availability
    Log in for availability
    1 * 100 pezzi
    1 * 100 pezzi - 43,40 €
    43,40 €
    1 * 100 pezzi
       43,40 €

  • 129-1619
    Cable ties, polyamide 6.6, resistance: 22
    Cable ties, polyamide 6.6, resistance: 22
    Blue
    200×4,8 mm
    Log in for availability
    Log in for availability
    1 * 100 pezzi
    1 * 100 pezzi - 24,20 €
    24,20 €
    1 * 100 pezzi
       24,20 €

  • 129-1620
    Cable ties, polyamide 6.6, resistance: 22
    Cable ties, polyamide 6.6, resistance: 22
    Green
    200×4,8 mm
    Log in for availability
    Log in for availability
    1 * 100 pezzi
    1 * 100 pezzi - 24,20 €
    24,20 €
    1 * 100 pezzi
       24,20 €

  • 129-1621
    Cable ties, polyamide 6.6, resistance: 22
    Cable ties, polyamide 6.6, resistance: 22
    Orange
    200×4,8 mm
    Log in for availability
    Log in for availability
    1 * 100 pezzi
    1 * 100 pezzi - 24,20 €
    24,20 €
    1 * 100 pezzi
       24,20 €

  • 129-1617
    Cable ties, polyamide 6.6, resistance: 8
    Cable ties, polyamide 6.6, resistance: 8
    Red
    100×2,5 mm
    Log in for availability
    Log in for availability
    1 * 100 pezzi
    1 * 100 pezzi - 15,30 €
    15,30 €
    1 * 100 pezzi
       15,30 €

  • 129-1618
    Cable ties, polyamide 6.6, resistance: 22
    Cable ties, polyamide 6.6, resistance: 22
    Red
    200×4,8 mm
    Log in for availability
    Log in for availability
    1 * 100 pezzi
    1 * 100 pezzi - 24,20 €
    24,20 €
    1 * 100 pezzi
       24,20 €

  • 129-1612
    Cable ties, polyamide 6.6, resistance: 8
    Cable ties, polyamide 6.6, resistance: 8
    White
    100×2,5 mm
    Log in for availability
    Log in for availability
    1 * 100 pezzi
    1 * 100 pezzi - 14,70 €
    14,70 €
    1 * 100 pezzi
       14,70 €

  • 129-1613
    Cable ties, polyamide 6.6, resistance: 18
    Cable ties, polyamide 6.6, resistance: 18
    White
    150×3,6 mm
    Log in for availability
    Log in for availability
    1 * 100 pezzi
    1 * 100 pezzi - 18,20 €
    18,20 €
    1 * 100 pezzi
       18,20 €

  • 129-1614
    Cable ties, polyamide 6.6, resistance: 22
    Cable ties, polyamide 6.6, resistance: 22
    White
    200×4,8 mm
    Log in for availability
    Log in for availability
    1 * 100 pezzi
    1 * 100 pezzi - 22,40 €
    22,40 €
    1 * 100 pezzi
       22,40 €

  • 129-1615
    Cable ties, polyamide 6.6, resistance: 22
    Cable ties, polyamide 6.6, resistance: 22
    White
    300×4,8 mm
    Log in for availability
    Log in for availability
    1 * 100 pezzi
    1 * 100 pezzi - 37,70 €
    37,70 €
    1 * 100 pezzi
       37,70 €

  • 129-1616
    Cable ties, polyamide 6.6, resistance: 22
    Cable ties, polyamide 6.6, resistance: 22
    White
    370×4,8 mm
    Log in for availability
    Log in for availability
    1 * 100 pezzi
    1 * 100 pezzi - 43,40 €
    43,40 €
    1 * 100 pezzi
       43,40 €

  • 129-1622
    Cable ties, polyamide 6.6, resistance: 22
    Cable ties, polyamide 6.6, resistance: 22
    Yellow
    200×4,8 mm
    Log in for availability
    Log in for availability
    1 * 100 pezzi
    1 * 100 pezzi - 24,20 €
    24,20 €
    1 * 100 pezzi
       24,20 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.