Order Entry
Italy
Orders LinkContactUsLinkComponent
SeramunBlock
SeramunBlock
Supplier:   Apollo Scientific
SeramunBlock
Supplier:  Apollo Scientific
Certificates
3 Options Available Below
Product Details & Documents
Combined Stabiliser/ blocking solution for solid phase immobilised proteins effecting protein stabilisation and blocking of free surfaces.
Product Family Options
Product Information
  • Pk
    Availability
    Price
  • SeramunBlock
    100 mL
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL - 82,50 €
    82,50 €
    1 * 100 mL
       82,50 €
  • SeramunBlock
    250 mL
    Log in for availability
    Log in for availability
    1 * 250 mL
    1 * 250 mL - 98,40 €
    98,40 €
    1 * 250 mL
       98,40 €
  • APOSBIL3122-5L
    SeramunBlock
    SeramunBlock
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 747,00 €
    747,00 €
    1 * 5 L
       747,00 €
    Specifications:
    New Product