Recombinant keratinase
Catalog # APOSBIPR1001-500MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pk:500 mg
- Enzyme type:Recombinant
- Storage conditions:Freezer
- Enzyme name:Keratinase
Specifications
About this item
Activity > 12 U/mg. from Bacillus licheniformis and produced in E. Coli and B. Megaterium. Endotoxin negative. Sterile lyophilized powder.