Candida rugosa Lipase
Supplier: Apollo Scientific
Certificates
About this item
Covalent on IB-150A. Activity: 250 TBU/g
- immobilized on Immobead 150
Specifications
- Species:Candida rugosa
- Storage conditions:Refrigerator
- Enzyme name:Lipase
Specifications
Frequently Bought Together