Order Entry
Italy
ContactUsLinkComponent
Clostridium kluyveri Diaphorase
Clostridium kluyveri Diaphorase
Catalog # APOSBIB3058-10KU
Supplier:  Apollo Scientific
CAS Number:  
Clostridium kluyveri Diaphorase
Catalog # APOSBIB3058-10KU
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pk:
    1
  • Species:
    Clostridium kluyveri
  • Storage conditions:
    Freezer
  • MDL No.:
    MFCD00130947
  • Enzyme name:
    Diaphorase
  • Enzyme synonyms:
    NAD(P)H dehydrogenase (quinone), NAD(P)H: (quinone-acceptor) oxidoreductase
  • Pk Info:
    10 KU

Specifications

About this item

Lyophilized powder. From Clostridium kluyveri. Specific activity: IU/mg powder>20 units. IU per 1mg powder is approximately 300 units.