Recombinant HIV-Ip24 (from E. coli)
Catalog # APOSBITP3208-100UG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pk:100 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Protein/peptide name:HIV-Ip24
Specifications
About this item
Recombinant HIV-I produced in E. coli, 1mg/mg, >95% (HPLC, PAGE)