Order Entry
Italy
ContactUsLinkComponent
 
Recombinant Hepatitis C Virus NS5, Biotin
 
Recombinant Hepatitis C Virus NS5, Biotin

 

  • Conjugation:
    Biotin
  • Protein/peptide type:
    Recombinant
  • Protein synonyms:
    HCV NS5
  • Protein/peptide name:
    Hepatitis C Virus NS5

 

Frequently Bought Together