Recombinant SARS Associated Coronavirus Nucleocaspid (1-49)
Catalog # APOSBITP1729-1MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pk:1 mg
- Protein/peptide type:Recombinant
- Protein synonyms:SARS Nucleocaspid
- Protein/peptide name:SARS Associated Coronavirus Nucleocaspid (1-49)
- Sequence:1-49