Recombinant SARS Associated Spike Mosaic S(N)
Catalog # APOSBITP1725-1MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pk:1 mg
- Protein/peptide type:Recombinant
- Protein synonyms:SARS S(N)
- Protein/peptide name:SARS Associated Spike Mosaic S(N)