Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Proteins and Peptides
Human recombinant cardiac Troponin-C
Human recombinant cardiac Troponin-C
APOSBITP1674-1MG
:
Apollo Scientific
:
Human recombinant cardiac Troponin-C
APOSBITP1674-1MG
:
Apollo Scientific
:
Pk:
1 mg
Protein/peptide type:
Recombinant
Species:
Human
Protein synonyms:
Troponin-C
Protein/peptide name:
Cardiac Troponin-C