Order Entry
Italy
ContactUsLinkComponent
Human recombinant Fetuin-B
Human recombinant Fetuin-B
Catalog # APOSBITP1557-2UG
Supplier:  Apollo Scientific
CAS Number:  
Human recombinant Fetuin-B
Catalog # APOSBITP1557-2UG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pk:
    2 g
  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Seq:
    02
  • Protein synonyms:
    Fetuin
  • Protein/peptide name:
    Fetuin-B

Specifications

About this item