Human Recombinant ERK1/MAPK3 Active Enzyme
Catalog # APOSBITP1400-20UG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pk:20 µG
- Protein/peptide type:Recombinant
- Species:Human
- Protein synonyms:ERK1/MAPK3
- Protein/peptide name:ERK1/MAPK3 Active Enzyme