Order Entry
Italy
ContactUsLinkComponent
 
Human recombinant HGF (from CHO cells)
 
Human recombinant HGF (from CHO cells)

 

  • Protein/peptide type:
    Recombinant
  • Source:
    CHO cells
  • Species:
    Human
  • Protein synonyms:
    Hepatocyte Growth Factor
  • Protein/peptide name:
    HGF

 

Frequently Bought Together