Human recombinant GM-CSF (from P. pastoris)
Supplier: Apollo Scientific
Certificates
About this item
Specifications
- Protein/peptide type:Recombinant
- Source:P. pastoris
- Species:Human
- Protein synonyms:Granulocyte Macrophage-Colony Stimulating Factor
- Protein/peptide name:GM-CSF
Specifications
Frequently Bought Together