Human recombinant MIP-1a
Catalog # APOSBITC2033-1MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pk:1 mg
- Protein/peptide type:Recombinant
- Species:Human
- Protein synonyms:Macrophage Inflammatory protein-1 alpha, CCL3
- Protein/peptide name:MIP-1a