Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Herboxidiene
Herboxidiene ≥97%
Herboxidiene ≥97%
Irritant
Herboxidiene ≥97%
Catalog # APOSBIBR1156-1MG
Supplier:
Apollo Scientific
CAS Number:
Expand image
Irritant
Herboxidiene ≥97%
Catalog # APOSBIBR1156-1MG
Supplier:
Apollo Scientific
CAS Number:
Specifications
Pack type:
Bottle
Pk:
1 mg
Seq:
2070
Specifications
About this item
Product Details
:
:
GHS07
Storage Temperature:
Freezer
CAS Number:
142861-00-5
Product Details