Order Entry
Italy
Orders LinkContactUsLinkComponent
Calcein

Calcein

Supplier:   Apollo Scientific
Calcein
Supplier:  Apollo Scientific
Certificates

2 Options Available Below

Product Details & Documents
Synonyms:  Oftasceina, Fluorescein Complexon, Oftasceinum, 2,7-Bis[bis(carboxymethyl)aminomethyl]fluorescein, Oftasceine, 2,2',2'',2'''-(((3',6'-Dihydroxy-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-2',7'-diyl)bis(methylene))bis(azanetriyl))tetraacetic acid, Fluorexon, Bis[N,N-bis(carboxymethyl)aminomethyl]fluorescein, NSC 298193
  • CAS Number:154071-48-4
  • Formula:C₃₀H₂₆N₂O₁₃
  • MDL Number:MFCD00005049
  • Molecular Weight:622.53 g/mol
  • Storage Temperature:Ambient
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 57,90 €
    57,90 €
    1 * 1 g
       57,90 €

  • Bottle
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 138,00 €
    138,00 €
    1 * 5 g
       138,00 €