About this item
Photometric test kits for the determination of formaldehyde in water samples.
- Measurement directly in the test tube, saving time and reducing exposure to chemical reagents
- Bar code cuvette identification for easy test call up in NANOCOLOR® spectrophotometers
- Precisely pre-dosed reagents with constant high quality from batch to batch
- Colored pictogram instructions and easy handling of the tubes
- 16 mm (OD) tubes for easy pipetting
The test kits are evaluated in 16 mm (AD) round cuvettes. They are pre-programmed on NANOCOLOR® photometers and are also compatible with METTLER TOLEDO easy VIS and easy UV. Programming data for other spectrophotometers are available.
Sludge reagent set: No
Digestion set: No
NanOx Metal: No
NanOx N: No
Digestion temperature: 60 °C / 140 °F (for MANA985046)
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Model:MANA985046
- Platform:NANOCOLOR® tube tests
- Storage temperature:15…25 °C
- Shelf life (months):24
- Weight:407,3 g (including packaging)
Specifications
Related Information
Certificates
Search by Lot NumberIFU NANOCOLOR® Photometric Tube Tests, Formaldehyde.pdf IFU Photometric tube tests, NANOCOLOR® Formaldehyde 10.pdf
Manual NANOCOLOR® Photometric Tube Tests, Formaldehyde 8.pdf Manual Photometric tube tests, NANOCOLOR® Formaldehyde 10.pdf