

4-(Trifluoromethylthio)phenyl isocyanate 97%
Catalog # APOSPC9864-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:219.19 g/mol
Boiling Pt:92…94 °C/15mmHg
Density:1.385 g/cm³ (25 °C)
Flash Pt:80 °C
UN:2206
ADR:6.1,III
:GHS06
:GHS07
Storage Temperature:Refrigerator
CAS Number:24032-84-6
Formula:C₈H₄F₃NOS
MDL Number:MFCD00036025