
2,2-Bis(3,5-difluoro-4-hydroxyphenyl)hexafluoropropane 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:114611-30-2
:
Formula:C₁₅H₆F₁₀O₂
MDL Number:MFCD01320803
:408.2 g/mol
Melting Pt:156…160 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together