Order Entry
Italy
ContactUsLinkComponent
2-[3-(Trifluoromethyl)phenyl]-1,3-thiazole-4-carboxylic acid
Irritant
2-[3-(Trifluoromethyl)phenyl]-1,3-thiazole-4-carboxylic acid
Catalog # APOSPC9506-50MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2-[3-(Trifluoromethyl)phenyl]-1,3-thiazole-4-carboxylic acid
Catalog # APOSPC9506-50MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    50 mg
  • Seq:
    2370

Specifications

About this item

 :
 :273.24 g/mol
Melting Pt:200…202 °C
 :GHS07
Storage Temperature:Refrigerator
CAS Number:115311-32-5
Formula:C₁₁H₆F₃NO₂S
MDL Number:MFCD00142039