Order Entry
Italy
ContactUsLinkComponent
6-Methyl-2-oxo-4-(trifluoromethyl)-1,2-dihydro-3-pyridinecarbonitrile
Irritant
6-Methyl-2-oxo-4-(trifluoromethyl)-1,2-dihydro-3-pyridinecarbonitrile
Catalog # APOSPC9432-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
6-Methyl-2-oxo-4-(trifluoromethyl)-1,2-dihydro-3-pyridinecarbonitrile
Catalog # APOSPC9432-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :202.14 g/mol
Melting Pt:232…234 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:654-49-9
Formula:C₈H₅F₃N₂O
MDL Number:MFCD01314908