
Methyl-3-{[3-chloro-5-(trifluoromethyl)-2-pyridinyl]oxy}-2-thiophenecarboxylate 97%
Catalog # APOSPC9402-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
Melting Pt:63…64 °C
:337.71 g/mol
:GHS07
Storage Temperature:Refrigerator
CAS Number:338756-06-2
Formula:C₁₂H₇ClF₃NO₃S
MDL Number:MFCD00141796